Learn More
Medchemexpress LLC Glycopyrrolate | 596-51-0 | 398.33 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1746510MM/1ML
Encompass_Preferred
Glycopyrrolate (Glycopyrronium bromide), a quaternary ammonium derivative, is a muscarinic receptor antagonist. It has a bronchoprotective effect and produces a beneficial effect on blood pressure. Glycopyrrolate can be used for the research of bronchial diseases.
  • Muscarinic receptor antagonist
  • Bronchoprotective effect
  • Beneficial effect on blood pressure
  • Can be used for the research of bronchial diseases
Catalog No. 50-003-86433