Learn More
Medchemexpress LLC Glycopyrrolate | 596-51-0 | 99.9% | 398.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY174655MG
Encompass_Preferred
Glycopyrrolate (Glycopyrronium bromide) is a quaternary ammonium derivative and a muscarinic receptor antagonist. It has a bronchoprotective effect and positively influences blood pressure. It is used in research for bronchial diseases.
  • Muscarinic receptor antagonist
  • Bronchoprotective effect
  • Beneficial effect on blood pressure
  • Used for research of bronchial diseases
Catalog No. 50-003-86707