Learn More
Waters Corp GlycoWorks HILIC Flangeless Cartridge, 1 cc, 20 Pack
Supplier:  Waters Corp 186007239
Encompass_Preferred
This pack contains 20 single-use, 1 cc flangeless cartridges, ideal for glycan extraction and cleanup when using a positive pressure manifold (PPM). They effectively remove contaminants like salts and detergents from hydrophilic analytes prior to MS analysis and eliminate excess labeling reagent. Each cartridge holds 10 mg of sorbent, supporting 10 reactions with 2-AB or 20 with RapiFluor-MS.
  • Single-use, 1 cc flangeless format
  • Compatible with positive pressure manifold (PPM) systems
  • Extracts and cleans up free or FLR labeled glycans
  • Removes contaminants (salts, detergents) from hydrophilic analytes before MS
  • Eliminates excess labeling reagent
  • Each cartridge contains 10 mg of sorbent
  • Supports 10 reactions (2-AB) or 20 reactions (RapiFluor-MS)
  • Supplied as a 20-pack
  • Store dry at room temperature, 1-year minimum shelf life (unopened)
Catalog No. 50-239-2731