Learn More
Waters Corp Waters GlycoWorks Rapid Deglycosylation Kit, 96 Samples, Sample Preparation, HILIC, MS Detection
Supplier:  Waters Corp 186008840
Encompass_Preferred
The GlycoWorks Rapid Deglycosylation Kit provides a streamlined workflow for glycan characterization, enabling rapid quantification and validation of deglycosylation efficiency. It supports the comparison of intact with partial PNGase F deglycosylation for site occupancy and intact O-linked analysis, facilitating faster sample preparation, data collection, and interpretation.
  • Allows for rapid quantification and validation of deglycosylation efficiency.
  • Reduces sample preparation time to less than one hour.
  • Provides increased fluorescence quantification and superior mass spectral response.
  • Simplifies complex sample preparation and data interpretation.
  • Supports 96 sample protocol for N-glycan analysis.
  • Improves in-solution protein digestion speed and peptide recovery.
  • Utilizes HILIC separation mode.
  • Compatible with MS, MS/MS, UV, HRMS detection methods.
Catalog No. 50-146-1887