Learn More
Waters Corp Glycoworks Sample Collection Module
Supplier:  Waters Corp 186007988
Encompass_Preferred
The GlycoWorks Sample Collection Module contains all needed plastics to properly handle all samples and run the SPE for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow. The kit contains the following: (15) strips of 8-1mL polypropylene tubes, (1) 96-Well Rack for 1 ml tubes, (15) Strips of 8 Plug Caps for 1 ml tubes, (15) strips of 8 600ul tubes, (15) strips of 8 Pre-slit Silicone/PTFE Cap Mats, (1) 1ml Round Collection Plate and (1) Waste Tray.
Catalog No. NC1675416