Learn More
Waters Corp WATERS CORPORATION GlycoWorks SPE Reagents, Automation, 1 pk
Supplier: Waters Corp 186008747
The GlycoWorks SPE Reagents are designed for the solid-phase extraction (SPE) sample cleanup portion of the GlycoWorks RapiFluor-MS N-Glycan Kit. These reagents are scaled to residual volumes needed for automation platforms.
- Contains all needed reagents for SPE sample cleanup.
- Scaled for automation platforms.
- Shipping condition is ambient.
- Product type is reagent.
- Units per package: 1 pack.
- Includes two 12 mL vials of elution buffer.
- Includes two 22 mL vials of sample diluent.
- Includes two 18 mL vials of equilibration solvent.
- Includes two 70 mL vials of wash solvent.
- This product contains hazardous material, which may require special freight handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.