Learn More
Cayman Chemical GlycRophsphoNEIcosapentae 10mg
SDP
Supplier:  Cayman Chemical 9000184210 MG
Encompass_Preferred
The metabolic precursor of EPEA, an N-acylethanolamide that inhibits dietary-restriction-induced lifespan extension in wild type and TOR pathway mutant nematodes; also has anti-inflammatory properties
Catalog No. 50-289-4151