Learn More
Medchemexpress LLC Glycyl-L-alanine | 3695-73-6 | MFCD00065112 | 97.0% | 146.14 g/mol | C5H10N2O3 | 5 G

Supplier: Medchemexpress LLC HYW1419105G
Gly-Ala (H-Gly-Ala-OH) is a dipeptide composed of glycine and L-alanine supplied as a white to off-white solid for research use. It is commonly used in peptide synthesis, biochemical assays, and as a simple model compound in studies of protein folding and enzymatic activity. Documentation for quality and safety is provided with each laboratory-scale package.
- Dipeptide of glycine and L-alanine.
- Molecular weight 146.14 g/mol.
- Purity 97.0% (supplier specification).
- White to off-white solid physical form.
- Available in small lab-scale sizes for research use.
- Provided with datasheet, SDS, and COA for quality and safety.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.