Learn More
Medchemexpress LLC Glycyl-L-glutamine | 13115-71-4 | MFCD00077124 | 98.0% | 203.20 g·mol⁻¹ | C7H13N3O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11754110MM 1ML
Encompass_Preferred
Glycyl-L-glutamine is a dipeptide composed of glycine and L-glutamine (CAS 13115-71-4) used in biochemical and neuropharmacological research. It is supplied as a ready-to-use 10 mM aqueous solution (1 mL) and is also available as a solid for custom preparation. Purity is typically 98% and the compound is water soluble at research concentrations.
  • acts as an antagonist of beta-endorphin(1-31) in some systems.
  • ready-to-use 10 mM aqueous solution provided for immediate experiments.
  • also available in solid form for formulation flexibility.
  • high purity suitable for research applications.
  • water soluble at typical research concentrations.
Catalog No. 50-002-27625