Learn More
Medchemexpress LLC Glycyl-L-glutamine | 13115-71-4 | MFCD00077124 | 98.0% | 203.20 g/mol | C7H13N3O4 | 100 MG

Supplier: Medchemexpress LLC HY117541100MG
Glycyl-L-glutamine is a research-grade dipeptide used in biochemical and cell-culture research as a stable glutamine-containing peptide and an enzymatic cleavage product of beta-endorphin with reported antagonist activity.
- Dipeptide of glycine and L-glutamine.
- CAS number 13115-71-4.
- Purity 98.0%.
- Water soluble; may require ultrasonic treatment for full dissolution.
- Stable powder with defined storage recommendations for long-term stability.
- Available in small research pack sizes such as 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.