Learn More
Medchemexpress LLC Glycyl-L-histidyl-L-lysine | 49557-75-7 | 99.9% | 340.38 | 10 G
SDP
Supplier:  Medchemexpress LLC HYP004610G
Encompass_Preferred
Glycyl-L-histidyl-L-lysine is a tripeptide composed of glycine, L-histidine, and L-lysine residues linked in sequence. It functions as a hepatotropic immunosuppressor and exhibits an anxiolytic effect. Glycyl-L-histidyl-L-lysine and its copper complexes are known for good skin tolerance.
  • Composed of glycine, L-histidine, and L-lysine residues.
  • Functions as a hepatotropic immunosuppressor.
  • Exhibits an anxiolytic effect.
  • Known for good skin tolerance with copper complexes.
  • Affects secretion of pro-inflammatory IL-6.
  • Decreases TNF-α-dependent IL-6 secretion.
  • Can stimulate mitotic activity of hepatocytes.
  • Can suppress immune reactivity dose-dependently.
Catalog No. 50-004-26211