Learn More
Medchemexpress LLC Glycyl-L-leucine | 869-19-2 | MFCD00008127 | 99.5% | 188.22 g/mol | C8H16N2O3 | 5 G

Supplier: Medchemexpress LLC HYW0160775G
Glycyl-L-leucine is a dipeptide composed of glycine and L-leucine used as a biochemical reagent and as a substrate in enzymatic assays. It is supplied at high purity for research applications and is suitable as an analytical standard or reagent for studying peptide metabolism.
- High purity (reported ~99.5%) suitable for analytical use.
- Serves as a substrate for glycyl-leucine dipeptidase assays.
- Molecular weight 188.22 g/mol; CAS 869-19-2.
- Available in multiple pack sizes for research needs.
- Soluble in water for convenient preparation of solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.