Learn More
Medchemexpress LLC Glycyl-L-leucine | 869-19-2 | MFCD00008127 | 99.5% | 188.22 g/mol | C8H16N2O3 | 50 G

Supplier: Medchemexpress LLC HYW01607750G
Glycyl-L-leucine is a dipeptide composed of glycine and L-leucine, used primarily as a substrate for glycyl-leucine dipeptidase and as a reference standard in biochemical and metabolite studies. Supplied as a high-purity powder for laboratory research; not for clinical or human use.
- High purity (≈99.5%).
- Molecular formula C8H16N2O3; molecular weight 188.22 g/mol.
- Useful as a substrate for enzymatic assays and as a reference standard.
- Available in multiple laboratory sizes, including 50 g.
- Certificate of analysis and datasheet available.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.