Learn More
Medchemexpress LLC Glycyl-L-phenylalanine | 3321-03-7 | MFCD00065110 | 99.7% | 222.24 g/mol | C11H14N2O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0139981G
Encompass_Preferred
Glycyl-L-phenylalanine is a dipeptide formed from glycine and L-phenylalanine. It is supplied as a high-purity research reagent suitable for use as an analytical standard, synthesis intermediate, and research tool in peptide chemistry and enzymology.
  • Dipeptide composed of glycine and L-phenylalanine.
  • High purity suitable for analytical use (reported 99.7%).
  • Molecular weight 222.24 g/mol; formula C11H14N2O3.
  • Suitable as a synthesis intermediate and reference standard.
  • White to off-white solid convenient for storage and handling.
Catalog No. 50-003-03813