Learn More
Medchemexpress LLC Glycyl-L-proline 7-amido-4-methylcoumarin hydrobromide | 115035-46-6 | MFCD00152092 | 98.7% | 410.26 g·mol⁻¹ | C17H20BrN3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1378345MG
Encompass_Preferred
Glycyl-L-proline 7-amido-4-methylcoumarin hydrobromide (CAS 115035-46-6) is a fluorogenic dipeptide substrate used to measure dipeptidyl peptidase IV (DPP-IV) activity. Enzymatic cleavage releases the fluorescent 7-amido-4-methylcoumarin (AMC) reporter, enabling sensitive continuous or end-point assays in biochemical and enzymatic studies. Supplied as an off-white to light yellow solid with high reported purity for research use.
  • Specific fluorescent substrate for detecting DPP-IV activity.
  • Releases 7-amido-4-methylcoumarin for sensitive fluorescence detection.
  • Suitable for continuous or end-point enzyme assays.
  • Compatible with standard fluorescence readers (abs/em ~353/442 nm).
  • Supplied as a solid with high reported purity for biochemical use.
Catalog No. 50-002-22973