Learn More
Medchemexpress LLC Glycyl-L-tryptophan | 2390-74-1 | MFCD00065978 | >98.0% | 261.28 g/mol | C13H15N3O3 | 100 MG

Supplier: Medchemexpress LLC HYW143241100MG
H-Gly-Trp-OH (Glycyl-L-tryptophan) is a research-grade dipeptide composed of glycine and L-tryptophan. Supplied as a dry powder for laboratory use, it is intended for peptide synthesis, biochemical assays, and metabolite or biochemical pathway studies. Handle and store according to supplier instructions to maintain stability.
- Composed of glycine and L-tryptophan.
- High purity (>98.0%).
- Molecular weight 261.28 g/mol.
- Molecular formula C13H15N3O3.
- Available in multiple sizes for research workflows.
- Powder storage: -20°C for long-term; follow supplier guidelines.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.