Learn More
Selleck Chemical LLC Glycyl-L-valine
SDP
Supplier:  Selleck Chemical LLC S612025MG
Encompass_Preferred
Glycyl-L-valine (H-Gly-Val-OH Glycylvaline) is a dipeptide composed of glycine and valine It is an incomplete breakdown product of protein digestion or protein catabolism
Catalog No. 50-000-04242