Learn More
Medchemexpress LLC Glycyl-prolyl-glutamic acid | 32302-76-4 | MFCD00144405 | 99.6% | 301.30 g/mol | C12H19N3O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117483100MG
Encompass_Preferred
Glycyl-prolyl-glutamic acid (Gly-Pro-Glu) is the N-terminal tripeptide of insulin-like growth factor-1 (IGF-1). It is a neuroactive peptide reported to modulate acetylcholine release and is supplied as a white to off-white solid for laboratory research use.
  • Neuroactive tripeptide that modulates acetylcholine release.
  • N-terminal tripeptide of insulin-like growth factor-1 (IGF-1).
  • High purity suitable for research applications (~99.6%).
  • Available as a solid in multiple pack sizes, including 100 mg.
  • Molecular weight 301.30 g/mol; formula C12H19N3O6.
  • For research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-28693