Learn More
Selleck Chemical LLC Glycylglycine S4810-25mg
SDP
Supplier:  Selleck Chemical LLC S481025MG
Encompass_Preferred
Glycylglycine (Diglycine Diglycocoll Glycine dipeptide) the simplest peptide made of two glycine molecules is usually used in the synthesis of more complicated peptides
Catalog No. 50-311-0132