Learn More
Alkali Scientific Glycylglycine [Gly Gly], 100 Grams
SDP
Supplier:  Alkali Scientific YCY002CP
Encompass
Glycylglycine (Gly Gly), a dipeptide composed of glycine and glycine, is provided in a 100g bottle for use in laboratory and cell culture applications. As a buffer component, Glycylglycine stabilizes pH in various biochemical reactions and helps maintain the desired conditions for cell growth. It is especially useful in protein purification, enzyme assays, and metabolic studies where the buffering capacity of a solution is crucial. Additionally, Glycylglycine is used in protein crystallization, receptor studies, and to study peptide activity in cell signaling. In cell culture, it serves to ensure optimal conditions for cell viability and function.
Catalog No. 50-006-61707