Learn More
Medchemexpress LLC Glycylglycine (standard) | 556-50-3 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD0889R50MG
Encompass_Preferred
Glycylglycine (Standard) is an analytical standard for research and analytical applications. It functions as a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor, contributing to energy metabolism and antioxidant processes. This compound supports spermatogonial stem cells (SSCs) proliferation and nerve myelin repair by upregulating neurotrophic factors. It is a reference standard for HPLC, GC, and MS.
  • Analytical standard for glycylglycine.
  • Non-selective glycylglycine dipeptidase substrate and iNOS inhibitor.
  • Participates in energy metabolism and antioxidant processes.
  • Supports spermatogonial stem cells (SSCs) proliferation.
  • Upregulates neurotrophic factors to support nerve myelin repair.
  • Reference standard for HPLC, GC, and MS research.
Catalog No. 50-004-10177