Learn More
Cayman Chemical Glycyrhza glabra Acarbose 10g
SDP
Supplier:  Cayman Chemical 1188510 G
Encompass_Preferred
An inhibitor of α-glucosidases (IC50s = 0.16 and 2.9 µM for maltase and sucrase, respectively); activates phosphorylase kinase in a cell-free assay at 250 µM; decreases urine levels of glucose and glycosylated hemoglobin, as well as glomerular mesangial thickening in a db/db mouse model of diabetic nephropathy at 40 mg/kg in the diet; reduces sucrose-induced increases in infarct size as a percentage of the area at risk and serum MDA levels in a mouse model of coronary artery ligation-induced cardiac ischemia-reperfusion injury at 10 mg/kg
Catalog No. 50-295-1650