Learn More
Medchemexpress LLC GlyH-101 | 328541-79-3 | 99.7% | 493.15 | 25 MG
Supplier: Medchemexpress LLC 25MGHY18336
GlyH-101 is a potent and reversible inhibitor of CFTR (Cystic Fibrosis Transmembrane conductance Regulator) and VSORC (Volume-Sensitive Outwardly Rectifying Chloride) conductance. It also exhibits antiproliferative activity by inhibiting cell growth in a dose-dependent manner. This compound is suitable for various in vitro and in vivo studies related to cystic fibrosis and other conditions involving chloride channel regulation.
- Potent CFTR inhibitor
- Reversible inhibitor of VSORC conductance
- Exhibits antiproliferative activity in PCT and PS120 cells
- Reduces cholera toxin-induced intestinal fluid secretion in mice
- Soluble in DMSO for in vitro applications
- Available in various solid quantities