Learn More
Agilent Technologies AdvanceBio APTS Human IgG N-glycan library
Supplier:  Agilent Technologies GKSP005
Encompass
Library consists of complex biantennary oligosaccharides consistent with N-glycans on normal human IgG. The reducing termini are derivatized with the fluorescent dye, 8-aminopyrene-1,3,6-trisulfonic acid trisodium salt (APTS) for separation by capillary electrophoresis (CE).
Catalog No. NC1066782