Learn More
Agilent Technologies GlykoPrep cleanup module, 96-ct (formerly ProZyme). GlykoPrep N-glycan sample preparation is a cartridge based workflow. Module for cleanup of glycans from aqueous solution. Compatible with microplate liquid handling platforms. Designed for cleanup with the InstantDye labeling modules.
Supplier:  Agilent Technologies GS96CU
Encompass
GlykoPrep cleanup module, 96-ct (formerly ProZyme). GlykoPrep N-glycan sample preparation is a cartridge based workflow. Module for cleanup of glycans from aqueous solution. Compatible with microplate liquid handling platforms. Designed for cleanup with the InstantDye labeling modules.
Catalog No. NC0137961