Learn More
Medchemexpress LLC Glyoxalase I inhibitor 3 | 1415388-25-8 | 98.1% | 375.42 g/mol | C22H21N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1466555MG
Encompass_Preferred
Glyoxalase I inhibitor 3 is a potent small-molecule inhibitor of glyoxalase I (GLO1) with a reported IC50 of 0.011 μM. It is supplied as a research reagent in milligram quantities for biochemical and pharmacological studies. Key chemical properties include molecular formula C22H21N3O3, molecular weight 375.42 g/mol, and measured purity of approximately 98.06%.
  • Potent GLO1 inhibition with IC50 0.011 μM.
  • Suitable for in vitro biochemical and cellular research.
  • High chemical purity (~98.1%).
  • Available in milligram quantities for assay development.
  • Chemical formula C22H21N3O3; molecular weight 375.42 g/mol.
Catalog No. 50-002-22316