Learn More
Medchemexpress LLC Glyphosate | 1071-83-6 | ≥98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0863500MG
Encompass_Preferred
Glyphosate is a non-selective systemic biocide with broad-spectrum activity, derived from the amino acid glycine. It acts by inhibiting the enzymatic activity of 5-endopyruvylshikimate 3-phosphate synthase (EPSPS) in the shikimic acid pathway, preventing the synthesis of aromatic amino acids. This product is for research use only.
  • Non-selective systemic biocide
  • Broad-spectrum activity
  • Inhibits EPSPS in the shikimic acid pathway
  • Prevents synthesis of aromatic amino acids
  • Induces oxidative stress, neuroinflammation, and mitochondrial dysfunction
Catalog No. 50-004-08382