Learn More
Medchemexpress LLC Glyphosate-d2 | 2733532-11-9 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0863S1MG
Encompass_Preferred
Glyphosate-d2 is the deuterium labeled Glyphosate, an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway in plants, a metabolic pathway not found in animals, which is required for the synthesis of aromatic amino acids. This compound can be used as a tracer and as an internal standard for quantitative analysis, with deuterium substitution having the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Glyphosate
  • Herbicidal derivative of amino acid glycine
  • Targets and blocks the shikimate pathway in plants
  • Used as a tracer and internal standard for quantitative analysis
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08860