Learn More
Medchemexpress LLC Gm-60186 | 99.3% | 504.59 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY169229100MG
Encompass_Preferred
GM-60186 is a potent 5-HT receptor 2B (HTR2B) inhibitor with an IC50 of 257 nM. It demonstrates significant inhibition of colorectal cancer proliferation and migration, and has been shown to reduce tumor size and weight in xenograft mouse models.
  • Inhibits 5-HT2B receptor activity
  • Reduces colorectal cancer cell proliferation and migration
  • Effective in vivo against xenograft tumors
  • Available in various pack sizes for research needs
  • Detailed documentation for research support
Catalog No. 50-003-90896