Learn More
Medchemexpress LLC GM-CSF 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7811020UG
Encompass_Preferred
GM-CSF R alpha is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2 GM-CSF R alpha binds to the cytokine with high specificity and low affinity[1] GM-CSF R alpha binds to GM-CSF and induces cell differentiation[2] GM-CSF R alpha monoclonal antibody decreases GM-CSFR expression on GM-CSF-responsive cells and shows anti-inflammatory activity in rheumatoid arthritis (RA)[3] GM-CSF R alpha Protein Human (Biotinylated HEK293 His-Avi) is a biotinylated recombinant protein with a C-Terminal His label and a C-Terminal Avi label It consists of 298 amino acids (E23-G320) and is produced in HEK293 cells
Catalog No. 50-001-87413