Learn More
Medchemexpress LLC GM-CSF Feline M36I 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP794325UG
Encompass_Preferred
The GM-CSF protein acts as a potent cytokine that coordinates the growth and differentiation of hematopoietic precursor cells of different lineages including granulocytes macrophages eosinophils and erythrocytes Structurally GM-CSF exists as a monomer and its signaling is mediated through a dodecamer complex GM-CSF Protein Feline (M36I T56A K126N) is the recombinant GM-CSF protein expressed by E coli with tag free
Catalog No. 50-001-94639