Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GM-CSF protein, human (CHO) | >95.0% | 2UG
SDP

Supplier:  Medchemexpress LLC HYP70162UG

Encompass_Preferred

Recombinant human granulocyte-macrophage colony-stimulating factor (GM-CSF) produced in Chinese hamster ovary (CHO) cells and supplied lyophilized for research use. It is a hematopoietic growth factor for cell proliferation and functional assays, with high purity and potent activity in TF-1 cell assays.

  • Expressed in CHO cells, untagged.
  • Purity greater than 95% by reducing SDS-PAGE.
  • High biological activity; ED50 <0.2 ng/mL in TF-1 assays.
  • Observed glycosylated molecular weight ~17-32 kDa.
  • Lyophilized formulation with trehalose for stability.
  • Stable at -20°C; aliquot and store at -80°C for extended storage.

Catalog No. 50-001-76954


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.