Learn More
Medchemexpress LLC GM-CSF Mouse HEK29 500ug
SDP
Supplier:  Medchemexpress LLC HYP70623500UG
Encompass_Preferred
Recombinant mouse granulocyte-macrophage colony-stimulating factor (GM-CSF) produced in HEK293 cells with a C-terminal His tag. Supplied as a 500 μg vial, the protein is intended for in vitro cell differentiation, proliferation, and signaling assays involving the GM-CSF receptor. Reported properties include a 124-amino-acid sequence and an apparent molecular weight of 18-30 kDa (glycosylated form).
  • Expressed in HEK293 cells to provide mammalian glycosylation.
  • Contains a C-terminal His tag for detection and purification.
  • 124 amino acids; apparent molecular weight 18-30 kDa (glycosylated).
  • Supplied as 500 μg for preparative and assay use.
  • Stable when stored at -20°C; shelf life reported up to 2 years.
  • Suitable for cell differentiation, proliferation, and signaling assays.
Catalog No. 50-001-84567