Learn More
Medchemexpress LLC GM-CSF Pig 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7925650UG
Encompass_Preferred
The GM-CSF protein acts as a potent cytokine that coordinates the growth and differentiation of hematopoietic precursor cells of different lineages including granulocytes macrophages eosinophils and erythrocytes Structurally GM-CSF exists as a monomer and its signaling is mediated through a dodecamer complex GM-CSF Protein Pig is the recombinant Porcine-derived GM-CSF protein expressed by E coli with tag free
Catalog No. 50-001-94691