Learn More
Medchemexpress LLC GM-CSF Rhesus macaq 500ug
SDP
Supplier:  Medchemexpress LLC HYP7184500UG
Encompass_Preferred
Recombinant granulocyte-macrophage colony-stimulating factor (GM-CSF) from rhesus macaque, supplied lyophilized at 500 μg for research use. The protein stimulates proliferation, differentiation, and activation of granulocytes and macrophages and is provided with characterization data suitable for in vitro biological assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Demonstrated biological activity with ED50 <0.1 ng/mL in TF-1 cell proliferation assay.
  • Tag-free recombinant protein expressed in E. coli.
  • Stable lyophilized format for extended storage and easy reconstitution.
Catalog No. 50-001-83531