Learn More
Medchemexpress LLC GM-CSF Rhesus macaq 5ug

Supplier: Medchemexpress LLC HYP71845UG
Recombinant GM-CSF protein from rhesus macaque supplied lyophilized for research use. The protein is expressed in E. coli, shows >95% purity by reducing SDS-PAGE, and has a predicted molecular mass of approximately 14.4 kDa. The provided sequence corresponds to residues A18-E144. Follow the recommended storage and reconstitution instructions to maintain activity and stability.
- Recombinant protein expressed in E. coli.
- Greater than 95% purity by reducing SDS-PAGE.
- Predicted molecular mass ~14.4 kDa.
- Sequence corresponds to A18-E144 of the mature protein.
- Lyophilized formulation with buffered and trehalose options for stability.
- Store at -20°C; reconstitute at ≥100 μg/mL and aliquot for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.