Learn More
Medchemexpress LLC GMF-beta Mouse 50ug
SDP
Supplier:  Medchemexpress LLC HYP7280150UG
Encompass_Preferred
GMF- protein is a multifaceted regulatory factor that coordinates brain cell differentiation stimulates neuroregeneration and inhibits tumor cell proliferation Phosphorylation initiated by phorbol esters is critical for its activity suggesting a dynamic regulatory mechanism in response to external signals GMF-beta Protein Mouse is the recombinant mouse-derived GMF-beta protein expressed by E coli with tag free
Catalog No. 50-001-81777