Learn More
Pfanstiehl Inc L-Histidine Hydrochloride Monohydrate, High Purity, Low Endotoxin, Low Metals, EP, JP, ChP (H-117) - 100G GMP, Multi-compendial - CAS: 5934-29-2 Injectable Excipient Grade (L-Histidine HCl)
Supplier:  Pfanstiehl Inc H117RD
Encompass
Pfanstiehl L-Histidine Hydrochloride, High Purity, Low Endotoxin, Low Metals is a multi-compendial amino acid that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl L-Histidine HCl meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl L-Histidine HCl is often used to buffer therapeutic proteins and is available as USP (United States Pharmacopoeia), EP (Ph. Eur.) (European Pharmacopoeia), BP (British Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). Compendial, U.S.P. , cGMP, multicompendial
Catalog No. NC2953551