Learn More
ACROBiosystems GMP Human Laminin 521 Protein
Supplier: ACROBiosystems GMPLA5H24100UG
Laminin 521 were expressed in Descemet's membrane and corneal endothelium. This laminin isoform significantly enhanced the in vitro adhesion and proliferation, and differentiation of HCECs. Laminin 511-E8 consists of the C-terminal end of the alpha 5, b