Learn More
Selleck Chemical LLC GMX1778 (CHS828) 10mg 200484-11-3
SDP
Supplier:  Selleck Chemical LLC S811710MG
Encompass_Preferred
GMX1778 (CHS828) is a potent and specific inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) with IC50 and Kd of < 25 nM and 120 nM, respectively. GMX1778 induces programmed cell death with apoptotic features. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5676