Learn More
Cytiva GN-6 Metricel Membrane Disc Filter - 0.45 µm, 47 mm, Gridded (100/pkg)
Supplier:  Cytiva 63020
Encompass
The GN-6 membrane is certified for the microbiological analysis of potable, waste, process, and natural waters in accordance with the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.-For microbiological analysis-Mixed cellulose ethers-Easy bacterial quantificationIdeal for isolation and enumeration of total and fecal coliforms, Ecoli, fecal Streptococcus, fungi, and other heterotrophic bacteria.GN-6 grids have a unique dot grid pattern that provides easy quantification of bacterial colonies without growth inhibition or enhancement. GN-4 membranes dissolve completely using standard digestion procedures, and clear completely for minimal interference in fiber counting.Ordering information: GN-6 is available gamma irradiated, individually packaged (except filter model 60016) or nonsterile, bulk packed. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3267
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.