Learn More
ABclonal Technology GNAQ Rabbit mAb
SDP
Supplier:  ABclonal Technology A2111120UL
Encompass_Preferred
This locus encodes a guanine nucleotide-binding protein The encoded protein an alpha subunit in the Gq class couples a seven-transmembrane domain receptor to activation of phospolipase C-beta Mutations at this locus have been associated with problems in platelet activation and aggregation A related pseudogene exists on chromosome 2
Catalog No. 50-000-07354