Learn More
ABclonal Technology GNAQ Rabbit pAb
SDP
Supplier:  ABclonal Technology A14736500UL
Encompass_Preferred
This locus encodes a guanine nucleotide-binding protein. The encoded protein, an alpha subunit in the Gq class, couples a seven-transmembrane domain receptor to activation of phospolipase C-beta. Mutations at this locus have been associated with problems in platelet activation and aggregation. A related pseudogene exists on chromosome 2.
Catalog No. 50-001-06468