Learn More
Selleck Chemical LLC GNE-049
SDP
Supplier:  Selleck Chemical LLC S8625100MG
Encompass_Preferred
GNE-049 is a highly potent and selective inhibitor of CBP It exhibits an IC50 of 1 1 nM in TR-FRET assay GNE-049 also effectively inhibits BRET and BRD4 with IC50 values of 12 nM and 4200 nM respectively
Catalog No. 50-000-01515