Learn More
Medchemexpress LLC GNE-064 | 1997321-20-6 | 99.7% | 327.38 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY149026
Encompass_Preferred
GNE-064 is a selective, orally active, and highly soluble inhibitor of SMARCA4, SMARCA2, and PBRM1 bromodomains 5. It functions as a chemical probe for agent synthesis research.
  • Selective inhibitor of SMARCA4, SMARCA2, and PBRM1 bromodomains 5
  • Orally active and highly soluble
  • Inhibits SMARCA4 with an IC50 of 0.035 μM
  • Inhibits SMARCA2 with an EC50 of 0.10 μM
  • Suitable as a chemical probe for agent synthesis research
Catalog No. 50-003-63315