Learn More
Medchemexpress LLC GNE-0723 | 1883518-31-7 | MFCD30829912 | 99.9% | 467.78 g·mol⁻¹ | C16H8ClF6N5OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10833750MG
Encompass_Preferred
GNE-0723 is a brain-permeable positive allosteric modulator of the N-methyl-D-aspartate receptor (NMDAR) with high potency at GluN2A and characterized pharmacology useful for in-vitro and in-vivo neuroscience research.
  • Potent GluN2A activity (EC50 21 nM).
  • Lower activity at GluN2C and GluN2D (EC50 7.4 μM and 6.2 μM).
  • High purity suitable for biochemical and in vivo assays.
  • Solid form, white to yellow, with recommended cold storage for stability.
  • Intended for research into NMDAR modulation and neurological disease models.
Catalog No. 50-002-42702
Add to Cart
Request a Quote