Learn More
Medchemexpress LLC GNE-0946 | 1677667-24-1 | 98.1% | 460.79 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19774
Encompass_Preferred
GNE-0946 is described as a potent and selective RORγ (RORc) agonist with an EC50 value of 4 nM for HEK-293 cells. It is intended for research use only and not for sale to patients.
  • Potent and selective RORγ (RORc) agonist.
  • EC50 value of 4 nM for HEK-293 cell.
  • Exhibits inverse agonist activity at GAL4-fused human RORc, RORa, and RORb in HEK293T cells.
  • Shows antiinflammatory activity in human PBMC, inhibiting IL-17 production.
  • Available in solid and solution forms.
Catalog No. 50-004-02176