Learn More
Medchemexpress LLC GNE-207 | 2158266-58-9 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HY12002810MM 1ML
GNE-207 is a potent, selective small-molecule inhibitor of the CBP/p300 bromodomain supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for biochemical and epigenetic research applications, including in vitro bromodomain binding and selectivity studies.
- Ready-to-use 10 mM solution in DMSO, 1 mL
- High purity, approximately 99.9%
- Molecular formula C29H30N6O3 and molecular weight 510.59 g/mol
- Suitable for in vitro biochemical and epigenetic assays
- Store in solvent at -80°C for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.