Learn More
Medchemexpress LLC Gne-235 | 3036574-72-5 | 98.5% | 244.29 g·mol⁻¹ | C13H16N4O | 10 MG

Supplier: Medchemexpress LLC HY15028910MG
GNE-235 is a small-molecule probe that selectively targets the second bromodomain of PBRM1. It binds the bromodomain with a reported KD of 0.28 ± 0.02 μM and is intended for cellular evaluation of PBRM1 function. The compound is supplied as a solid with documented purity, solubility, and storage recommendations for research use.
- Selective for the second bromodomain of PBRM1.
- Reported KD 0.28 ± 0.02 μM for target binding.
- Molecular weight 244.29 g·mol⁻¹; formula C13H16N4O.
- Purity typically 98.45% as supplied.
- Soluble in DMSO at ~100 mg/mL; ultrasonic agitation recommended.
- Powder storage at -20°C for long-term stability; in solvent store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.