Learn More
Medchemexpress LLC GNE-235 | 3036574-72-5 | C13H16N4O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15028910MM/1ML
Encompass_Preferred
GNE-235 is a compound that selectively targets the second bromodomain of PBRM1, exhibiting a KD of 0.28 ± 0.02 μM. It is utilized for evaluating the cellular function of PBRM1.
Catalog No. 50-003-74774